Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate, Reagents

Catalog No. 01337210
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-210 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-210 Supplier Reagents Holdings Llc Supplier No. C22200504A
Add to Cart
Add to Cart

Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kjeldahl Solution, 50% Sodium Hydroxide/2.5% Sodium Thiosulfate
Melting Point -7°C
Boiling Point 102°C
Color Colorless
CAS 10102-17-7, 1310-73-2, 7732-18-5
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Synonym Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.