Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Kjeldahl Tablets
SDP

Catalog No. 05405667 Shop All BUCHI Products
Change view
Click to view available options
Chemical Name or Material:
Antifoam
Missouri
Titanium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
05-405-667 Missouri
05-405-668 Antifoam
05-405-666 Titanium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-667 Supplier BUCHI Supplier No. 11072629
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Digestion aids for nitrogen determination.

  • Environmentally friendly: free of toxic mercury and selenium
  • Different tablets to fit individual needs
  • For digestion

Specifications

Chemical Name or Material Missouri
Melting Point 1067°C (Main Component)
Color Blue
CAS 7758-99-8, 7778-80-5
Quantity 1000 Pk.
Packaging Plastic Container
Percent Purity Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.