Learn More
BUCHI Kjeldahl Weighing Boats
SDP
Catalog No. 05405670 Shop All BUCHI Products
Change view
Click to view available options
Dimensions (L x W x H):
58 x 10 x 10 mm
Catalog No. Dimensions (L x W x H)
05-405-670 58 x 10 x 10 mm
1 options
Catalog No. 05-405-670 Supplier BUCHI Supplier No. 11060522
Add to Cart
Add to Cart
Constructed of nitrogen free paper.
  • Easy handling and transfer of sticky samples and powders
  • 58 x 10 x 10 mm
Specifications
Type Weighing Boat
For Use With (Application) Handling/Transfer of Sticky Samples and Powders
Dimensions (L x W x H) 58 x 10 x 10 mm