Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeltabs CT (Copper and Titanium) - 5, Foss™

Catalog No. 12053105
Change view
Click to view available options
Quantity:
1000 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-053-105 1000 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-053-105 Supplier Foss North America Supplier No. 60046514

missing translation for 'validMainframeMsg'

Composition per tablet: 5g K2SO4 + 0.15g CuSO4·5H2O + 0.15g TiO2

Specifications

Chemical Name or Material Copper and Titanium Kjeltabs
CAS 7758-99-8, 13463-67-7
Physical Form Flat Round Speckled Tablets
Quantity 1000 Tablets
Packaging 5g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.