Learn More
Velp™ KjTabs VCT - Kjeldahl Catalyst Tablets, Velp™

Description
- The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
- Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with genuine catalyst tablets.
- The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
- KjTabs™ VCM consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) as metal salt to accelerate the mineralization process.
- Universally applicable, with a low copper content 3.5g K2SO4 + 0.1g CuSO4 x 5H2O 1000 pcs.
- Most suitable choice when performing Kjeldahl analysis in a great variety of industries such as food and beverage, agriculture, environmental, chemical and petrochemical:
- Protein determination in Nuts and Nut Products (almonds, nuts, hazelnuts, coconuts), grains and cereals
- N/Protein determination in flour (soya beans and lupins)
- Crude protein determination in meat and meat products
- Protein determination in compressed and granular yeast
- Protein determination in sugar, syrup, molasses
- Protein determination in wheat spaghetti and macaroni, egg pasta
- Nitrogen content determination in urea
- Total nitrogen determination in water
- Nitrogen content determination in raw natural rubber and rubber latex
Specifications
Specifications
| Chemical Name or Material | VCT |
| Quantity | 1000/Pk., 1000 Pc./Box |
| Color | White |
| Packaging | Metal Tin |
| Grade | Reagent |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.