Learn More
Velp™ KjTabs VS - Kjeldahl Antifoam Tablets, Velp™

Description
- Specific antifoaming tablets consisting of Potassium sulphate and Silicone.
- Antifoaming agents are fundamental to the good outcome of the Kjeldahl analysis since the production of foam tends to reduce the original quantity of the sample during the initial phase of the mineralization process. This can occur in many agricultural samples or fat-rich products.
- VS Antifoam used for foam reduction 0.97 g K2SO4 + 0.03 g Silicone, 1000 pcs.
- Added in combination with KjTabs™ catalylst tablets.
- Most suitable choice when performing Kjeldahl analysis in a great variety of industries.
Specifications
Specifications
| Chemical Name or Material | VS |
| Color | White |
| Quantity | 1000/Pk., 1000 Pc./Box |
| Packaging | Metal Tin |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.