Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ KjTabs VST - Kjeldahl Catalyst Tablets, Velp™
SDP

Catalog No. 11392071
Change view
Click to view available options
Quantity:
1000/Pk., 1000 Pc./Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-392-071 1000/Pk., 1000 Pc./Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-071 Supplier Velp™ Supplier No. A00000277
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KjTabsTM VST Kjeldahl Catalyst Tablets are ideal for oily and fatty samples. The perfect solutions to speed up the digestion process.

  • The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
  • Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with catalyst tablets.
  • The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
  • KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
  • VTCT for tougher samples & even faster digestion (a scaled down version of VCT) 3.5g K2SO4+ 0.105g CuSO4x 5 H2O + 0.105g TiO21000 pcs
  • Most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.

Specifications

Chemical Name or Material VST
Quantity 1000/Pk., 1000 Pc./Box
Color White
Packaging Metal Tin
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.