Learn More
Velp™ KjTabs VTCT - Kjeldahl Catalyst Tablets, Velp™

Description
- The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
- Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with catalyst tablets.
- The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
- KjTabs™ VTCT consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) and Titanium (Ti) as metal salts to accelerate the mineralization process.
- VTCT for tougher samples & even faster digestion (a scaled down version of VCT) 3.5g K2SO4+ 0.105g CuSO4x 5 H2O + 0.105g TiO21000 pcs
- Most suitable choice when performing Kjeldahl analysis for Nitrogen determination in milk and dairy products.
Specifications
Specifications
| Chemical Name or Material | VTCT |
| Color | White |
| Quantity | 1000/Pk., 1000 Pc./Box |
| Packaging | Metal Tin |
| Grade | Reagent |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.