Learn More
Medchemexpress LLC KKI-5 TFA | 99.3% | 887.90 | 10 MG
SDP
Catalog No. 5000425785
Encompass_Preferred
KKI-5 TFA is a specific inhibitor of tissue kallikrein. It has been shown to attenuate breast cancer cell invasion and is intended for research use only.
  • Specific inhibitor of tissue kallikrein
  • Attenuates breast cancer cell invasion
  • Target: Kallikrein
  • For research use only
Catalog No. 50-004-25785 Supplier Medchemexpress LLC Supplier No. HYP0237A10MG
KKI-5 TFA is a specific inhibitor of tissue kallikrein. It has been shown to attenuate breast cancer cell invasion and is intended for research use only.
  • Specific inhibitor of tissue kallikrein
  • Attenuates breast cancer cell invasion
  • Target: Kallikrein
  • For research use only