Learn More
TARGETMOL CHEMICALS INC KKII5 10MG
SDP
Catalog No. 502576292
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KKII5 is a potent inhibitor of the lipoxygenase LOX-1 with an IC50 of 19 uM.KKII5 inhibits lipid peroxidation and enhances the ability of the active centers of some compounds to receive electrons. purity: 97%
Catalog No. 50-257-6292 Supplier TARGETMOL CHEMICALS INC Supplier No. T7728810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KKII5 is a potent inhibitor of the lipoxygenase LOX-1 with an IC50 of 19 uM.KKII5 inhibits lipid peroxidation and enhances the ability of the active centers of some compounds to receive electrons. purity: 97%