Learn More
Medchemexpress LLC HY-101865 5mg Medchemexpress, KKL-10 CAS:952849-76-2 Purity:>98%
SDP
Catalog No. 501967899
Encompass_Preferred
Medchemexpress, HY-101865 5mg KKL-10 CAS:952849-76-2 KKL-10 is a small-molecule ribosome rescue inhibitor with broad-spectrum antimicrobial activity against bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7899 Supplier Medchemexpress LLC Supplier No. HY1018655MG
Medchemexpress, HY-101865 5mg KKL-10 CAS:952849-76-2 KKL-10 is a small-molecule ribosome rescue inhibitor with broad-spectrum antimicrobial activity against bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.