Learn More
Selleck Chemical LLC KKL-35
SDP
Catalog No. 5000002923
Encompass_Preferred
KKL-35 inhibits the trans-translation tagging reaction with an IC50 of 0 9 M It has broad-spectrum antibiotic activity
Catalog No. 50-000-02923 Supplier Selleck Chemical LLC Supplier No. S65622MG
KKL-35 inhibits the trans-translation tagging reaction with an IC50 of 0 9 M It has broad-spectrum antibiotic activity