Learn More
Medchemexpress LLC Kkl-35 | 865285-29-6 | 99.5% | 317.7 g/mol | C15H9ClFN3O2 | 1 ML
SDP
Catalog No. 5000247936
Encompass_Preferred
KKL-35 is a small-molecule inhibitor of the bacterial trans-translation tagging reaction, reported to inhibit trans-translation with an IC50 of 0.9 μM. It is provided for research use and is offered both as a 10 mM solution in DMSO and in multiple solid vial sizes for in vitro studies.
  • High reported potency: IC50 0.9 μM against trans-translation tagging reaction.
  • Available as 10 mM solution in DMSO for direct experimental use.
  • Also available in solid form across multiple vial sizes (mg scale).
  • High purity, typically ≥99.5% as reported by supplier.
  • Molecular weight 317.7 g/mol and formula C15H9ClFN3O2 for formulation planning.
  • Intended for research use only (not for human or clinical use).
Catalog No. 50-002-47936 Supplier Medchemexpress LLC Supplier No. HY10186610MM 1ML
KKL-35 is a small-molecule inhibitor of the bacterial trans-translation tagging reaction, reported to inhibit trans-translation with an IC50 of 0.9 μM. It is provided for research use and is offered both as a 10 mM solution in DMSO and in multiple solid vial sizes for in vitro studies.
  • High reported potency: IC50 0.9 μM against trans-translation tagging reaction.
  • Available as 10 mM solution in DMSO for direct experimental use.
  • Also available in solid form across multiple vial sizes (mg scale).
  • High purity, typically ≥99.5% as reported by supplier.
  • Molecular weight 317.7 g/mol and formula C15H9ClFN3O2 for formulation planning.
  • Intended for research use only (not for human or clinical use).