Learn More
Medchemexpress LLC KL-11743 | 1369452-53-8 | 99.4% | C30H30N6O3 | 50 MG
SDP
Catalog No. 5000354428
Encompass_Preferred
KL-11743 is a potent, orally active, and glucose-competitive inhibitor of class I glucose transporters (GLUT1, GLUT2, GLUT3, and GLUT4). It blocks glucose metabolism and can synergize with electron transport inhibitors to induce cell death. Additionally, KL-11743 induces the formation of disulfide bonds in actin cytoskeletal proteins, leading to cellular disulfidptosis.
  • Potent, orally active, and glucose-competitive inhibitor of class I glucose transporters
  • Blocks glucose metabolism and synergizes with electron transport inhibitors to induce cell death
  • Induces cellular disulfidptosis
  • Inhibits the growth of various cancer cell lines in vitro
  • Effective in decreasing tumor growth in animal models
Catalog No. 50-003-54428 Supplier Medchemexpress LLC Supplier No. HY14559750MG
KL-11743 is a potent, orally active, and glucose-competitive inhibitor of class I glucose transporters (GLUT1, GLUT2, GLUT3, and GLUT4). It blocks glucose metabolism and can synergize with electron transport inhibitors to induce cell death. Additionally, KL-11743 induces the formation of disulfide bonds in actin cytoskeletal proteins, leading to cellular disulfidptosis.
  • Potent, orally active, and glucose-competitive inhibitor of class I glucose transporters
  • Blocks glucose metabolism and synergizes with electron transport inhibitors to induce cell death
  • Induces cellular disulfidptosis
  • Inhibits the growth of various cancer cell lines in vitro
  • Effective in decreasing tumor growth in animal models