Learn More
Selleck Chemical LLC KL-11743-E0805-5MG
SDP
Catalog No. 502389973
Encompass_Preferred
KL-11743 is a potent glucose-competitive inhibitor of the class I glucose transporters with IC50s of 115 137 and 90 nM for GLUT1 GLUT2 and GLUT3
Catalog No. 50-238-9973 Supplier Selleck Chemical LLC Supplier No. E08055MG
KL-11743 is a potent glucose-competitive inhibitor of the class I glucose transporters with IC50s of 115 137 and 90 nM for GLUT1 GLUT2 and GLUT3