Learn More
TARGETMOL CHEMICALS INC KL-11743 5MG
SDP
Catalog No. 502578999
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KL-11743 specifically blocks glucose metabolism triggering an acute collapse in NADH pools and a striking accumulation of aspartate indicating a dramatic shift toward oxidative phosphorylation in the mitochondria. purity: 98%
Catalog No. 50-257-8999 Supplier TARGETMOL CHEMICALS INC Supplier No. T95585MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KL-11743 specifically blocks glucose metabolism triggering an acute collapse in NADH pools and a striking accumulation of aspartate indicating a dramatic shift toward oxidative phosphorylation in the mitochondria. purity: 98%