Learn More
MyBioSource Mouse ELISA Kits, Krebs von den Lungen-6 (KL-6)
SDP
Supplier:  MyBioSource MBS260234996STRIPWELLS
Encompass_Preferred
The MyBioSource Mouse KL-6 ELISA Kit quantitatively measures KL-6 in various mouse biological fluids using a sandwich enzyme immunoassay. Coated with a KL-6 specific antibody, the 96-well plate binds the KL-6, which is then detected with a biotinylated antibody and HRP-conjugated streptavidin, producing a color change measured at 450 nm.
Catalog No. NC2854750