Learn More
Coast KL10 Key Chain Light
Catalog No. 17010033
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
17-010-033 LED
1 options
Catalog No. 17-010-033 Supplier Coast Supplier No. 30771
Add to Cart
Add to Cart
The KL10 keychain flashlight comes in handy when you least expect to need light—delivering 100 lumens of fixed-beam flood lighting with easy twist-on/off function.
  • The all-aluminum body weighs only 1.3 ounces at just 2.5 inches long and is fitted with rubber gaskets for a solid, IP54 storm-proof build
  • Exceptionally portable, this mighty mini shines a 36-meter beam for up to 40 minutes on a single AAA alkaline battery
Specifications
Light Source LED
Voltage 1.5 V
Weight (English) 1.3 oz.
Material Aluminum
Product Type LED Task Light