Learn More
COAST Cutlery KL30R Rechargeable Flashlight With UV
Catalog No. 19030065
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
19-030-065 LED
1 options
Catalog No. 19-030-065 Supplier COAST Cutlery Supplier No. 31126
  • $25.40
    $24.65 / Each of 1
    Save $0.75
    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The KL30R is the next generation every day carry rechargeable flashlight. Designed to be the ultimate multi-tool of lights, it combines the functions of a flashlight, headlamp, area light, and UV detection light into one versatile and pocket-sized device.
  • Battery Charges Inside Light - COAST ZITHION™ non-ported rechargeable batteries are built to charge from within your COAST light using a COAST charging system.
  • Battery Charging Level Indicator - Check the status of your batteries charge level with our built-in charge level indicator. (Solid red) when charging, (solid green) when fully charged.
  • Built-In USB-C Port - With a built-in USB-C port, your battery can be charged using a USB-C cord. Some COAST models can be charged both inside or outside the light.
  • Dust Resistant - Take on any dirty work environment with this dust resistant light.
  • Rechargeable Battery - Power up on your terms with a recharging system that connects to standard outlets and ports.
  • Turbo Mode - The XP lights not only features standard light modes but also COAST's signature Turbo Mode for overpowered light in critical situations.
  • Two-Way Clip - Store it away or shine it forward. The Two-Way Clip enables gives you flexibility to stash the light in your pocket or clip it to the brim of your hat.
  • Water Resistance - Don't worry about the weather. COAST lights that have passed the IP54-level resistance tests are splash-resistant and built with durable materials that stand up to nature.
Specifications
Light Source LED
Voltage 3.7 V
Color Black
Product Type Rechargeable LED Flashlight