Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KLA seq acetate | 98.1% | 1876.47 (free base) | 1 MG
SDP

Catalog No. 5000398413
Encompass_Preferred

KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells, forming a delivery system that covalently links targeted mRNA and fluorescein. This system exhibits low re-export and is less resistant to serum effects.

  • Cell membrane-penetrating alpha-helical amphipathic model peptide
  • Conjugates peptide nucleic acids (PNAs) for mammalian cell delivery
  • Covalently links targeted mRNA and fluorescein
  • Low re-export and reduced serum effects
  • Purity: 98.1%
  • Molecular weight: 1876.47 (free base)
  • Appearance: Solid, white to off-white
  • Solubility: ≥ 100 mg/mL in H2O

Catalog No. 50-003-98413 Supplier Medchemexpress LLC Supplier No. HYP4084A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells, forming a delivery system that covalently links targeted mRNA and fluorescein. This system exhibits low re-export and is less resistant to serum effects.

  • Cell membrane-penetrating alpha-helical amphipathic model peptide
  • Conjugates peptide nucleic acids (PNAs) for mammalian cell delivery
  • Covalently links targeted mRNA and fluorescein
  • Low re-export and reduced serum effects
  • Purity: 98.1%
  • Molecular weight: 1876.47 (free base)
  • Appearance: Solid, white to off-white
  • Solubility: ≥ 100 mg/mL in H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.