Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KLA seq acetate | 98.1% | 1876.47 (free base) | 10 MG
SDP

Catalog No. 5000398412
Encompass_Preferred

KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells. It also covalently links targeted mRNA and fluorescein to form a delivery system. This system demonstrates low re-export and is less resistant to serum effects.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Lys-Leu-Ala-Leu-Lys-Leu-Ala-Leu-Lys-Ala-Leu-Lys-Ala-Ala-Leu-Lys-Leu-Ala-NH2
  • Storage: Sealed storage, away from moisture. Powder stored at -80°C for 2 years or -20°C for 1 year. In solvent, stored at -80°C for 6 months or -20°C for 1 month.
  • Solubility (in vitro): ≥ 100 mg/mL in H2O.

Catalog No. 50-003-98412 Supplier Medchemexpress LLC Supplier No. HYP4084A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells. It also covalently links targeted mRNA and fluorescein to form a delivery system. This system demonstrates low re-export and is less resistant to serum effects.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Lys-Leu-Ala-Leu-Lys-Leu-Ala-Leu-Lys-Ala-Leu-Lys-Ala-Ala-Leu-Lys-Leu-Ala-NH2
  • Storage: Sealed storage, away from moisture. Powder stored at -80°C for 2 years or -20°C for 1 year. In solvent, stored at -80°C for 6 months or -20°C for 1 month.
  • Solubility (in vitro): ≥ 100 mg/mL in H2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.