Learn More
Medchemexpress LLC KLA seq acetate | 98.1% | 1876.47 (free base) | 5 MG
SDP
Catalog No. 5000398414
Encompass_Preferred
KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells and covalently links targeted mRNA and fluorescein to form a delivery system with low re-export and less resistance to serum effects.
  • Cell membrane-penetrating α-helical amphipathic model peptide.
  • Conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells.
  • Forms a delivery system for targeted mRNA and fluorescein.
  • Exhibits low re-export and less susceptibility to serum effects.
Catalog No. 50-003-98414 Supplier Medchemexpress LLC Supplier No. HYP4084A5MG
KLA seq acetate is a cell membrane-penetrating α-helical amphipathic model peptide. It conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells and covalently links targeted mRNA and fluorescein to form a delivery system with low re-export and less resistance to serum effects.
  • Cell membrane-penetrating α-helical amphipathic model peptide.
  • Conjugates peptide nucleic acids (PNAs) for delivery to mammalian cells.
  • Forms a delivery system for targeted mRNA and fluorescein.
  • Exhibits low re-export and less susceptibility to serum effects.