Learn More
Aviva Systems Biology Corp KLC2 Antibody - C-terminal region (ARP62145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLC2. It was validated on Western Blot. . Alternate names: FLJ12387ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-587