Learn More
Aviva Systems Biology Corp KLC3 Antibody - middle region (ARP53345_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53345_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLC3. Alternate names: KLC2, KLC2L, KLCt, KNS2BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-61003