Learn More
Medchemexpress LLC KLD-12 TFA | 95.22% | 1467.79 | 5 MG
SDP
Catalog No. 5000422220
Encompass_Preferred
KLD-12 TFA is the TFA salt form of KLD-12, a 12-residue self-assembling peptide. It is used in tissue engineering and, when combined with SDF-1 self-assembled polypeptide, enhances chondrogenic differentiation of bone marrow stromal cells (BMSCs). The hydrogel form can fill full-thickness osteochondral defects in situ and improve cartilage repair. In vivo, it promotes bone regeneration at the fracture site in a dose-dependent manner.
  • Enhances chondrogenic differentiation of bone marrow stromal cells
  • Forms self-assembled polypeptide with SDF-1
  • Improves survival of bone marrow stromal cells
  • Promotes osteogenic differentiation of bone marrow stromal cells and cell migration via Wnt/β-catenin pathway
  • Promotes bone regeneration at the fracture site
Catalog No. 50-004-22220 Supplier Medchemexpress LLC Supplier No. HYP2263A5MG
KLD-12 TFA is the TFA salt form of KLD-12, a 12-residue self-assembling peptide. It is used in tissue engineering and, when combined with SDF-1 self-assembled polypeptide, enhances chondrogenic differentiation of bone marrow stromal cells (BMSCs). The hydrogel form can fill full-thickness osteochondral defects in situ and improve cartilage repair. In vivo, it promotes bone regeneration at the fracture site in a dose-dependent manner.
  • Enhances chondrogenic differentiation of bone marrow stromal cells
  • Forms self-assembled polypeptide with SDF-1
  • Improves survival of bone marrow stromal cells
  • Promotes osteogenic differentiation of bone marrow stromal cells and cell migration via Wnt/β-catenin pathway
  • Promotes bone regeneration at the fracture site