Learn More
MSC Klean-Strip Odorless Mineral Spirits, 1 gal
Catalog No. 501112397
Encompass
Klean-Strip Odorless Mineral Spirits is a highly refined solvent formulated for thinning oil-based paint, stain, and varnish. It features a very low odor and is composed of 100% hydrotreated light distillate (petroleum, mineral spirits), making it suitable for cleaning brushes and equipment with minimal residue.
  • Effective for thinning oil-based paint, stain, and varnish
  • Dries with minimal residue
  • Cleans brushes, tools, and equipment
  • Very low odor formulation
  • Suitable for outdoor applications
  • Supplied in a 1 gal. can
Catalog No. 50-111-2397 Supplier MSC Supplier No. 81332793
Klean-Strip Odorless Mineral Spirits is a highly refined solvent formulated for thinning oil-based paint, stain, and varnish. It features a very low odor and is composed of 100% hydrotreated light distillate (petroleum, mineral spirits), making it suitable for cleaning brushes and equipment with minimal residue.
  • Effective for thinning oil-based paint, stain, and varnish
  • Dries with minimal residue
  • Cleans brushes, tools, and equipment
  • Very low odor formulation
  • Suitable for outdoor applications
  • Supplied in a 1 gal. can