Learn More
MSC Klean-Strip Acetone, Liquid, Multi-Purpose Solvent, 5 gal
Catalog No. 501112393
Encompass
Klean-Strip Acetone is a strong, fast-acting solvent primarily used as a thinner and remover for specified coatings, including uncured polyester and epoxy resins, ink, adhesives, and contact cement. It also functions as an effective cleanup solvent for removing excess fiberglass resin or foreign material from solvent-tolerant surfaces after fiberglass projects, drying fast with no residue.
  • Strong, fast-acting solvent
  • Thinner and remover for specified coatings
  • Cleans project residue
  • Dries fast with no residue
  • Effective for cleaning up after fiberglass projects
Catalog No. 50-111-2393 Supplier MSC Supplier No. 70250337
Klean-Strip Acetone is a strong, fast-acting solvent primarily used as a thinner and remover for specified coatings, including uncured polyester and epoxy resins, ink, adhesives, and contact cement. It also functions as an effective cleanup solvent for removing excess fiberglass resin or foreign material from solvent-tolerant surfaces after fiberglass projects, drying fast with no residue.
  • Strong, fast-acting solvent
  • Thinner and remover for specified coatings
  • Cleans project residue
  • Dries fast with no residue
  • Effective for cleaning up after fiberglass projects