Learn More
ATCC Klebsiella aerogenes 6 x 0.3mL
Catalog No. 502383809
Encompass
ATCC 13048-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 13048™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-3809 Supplier ATCC Supplier No. 13048MINIPACK
ATCC 13048-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 13048™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.