Learn More
ATCC Klebsiella pneumoniae; 1000527, 7561
Catalog No. 502384298
Encompass
Klebsiella pneumoniae strain 10000527, 7561 was isolated from human urine. This strain was confirmed by PCR to be negative for blaKPC and positive for blaNDM. It has applications for multidrug testing and respiratory research.
Catalog No. 50-238-4298 Supplier ATCC Supplier No. BAA2146
Klebsiella pneumoniae strain 10000527, 7561 was isolated from human urine. This strain was confirmed by PCR to be negative for blaKPC and positive for blaNDM. It has applications for multidrug testing and respiratory research.