Learn More
ATCC Klebsiella pneumoniae; 6 x 0.3mL
Catalog No. 502383675
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain PCI 602 (ATCC 10031-MINI-PACK) consists of 6 ready-to-use vials of ATCC 10031 frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping. This product has applications in respiratory and infectious disease research, inhibition testing, and quality control.
Catalog No. 50-238-3675 Supplier ATCC Supplier No. 10031MINIPACK
Klebsiella pneumoniae subsp. pneumoniae strain PCI 602 (ATCC 10031-MINI-PACK) consists of 6 ready-to-use vials of ATCC 10031 frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping. This product has applications in respiratory and infectious disease research, inhibition testing, and quality control.