Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Klebsiella pneumoniae ATCC BAA-2814
SDP

Catalog No. 23047365 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-365 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-365 Supplier Microbiologics™ Supplier No. 01263L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal solution for many quality control applications including presence/absence testing, microbial identification methods, antimicrobial susceptibility testing, media QC, water testing and more

  • Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent, such as phosphate buffer or saline to inoculate culture media.
  • Packaged in a glass vial containing 6 pellets of a single strain

Specifications

ATCC® ATCC BAA-2814
Biosafety Level 2
Content And Storage 2°C to 8°C
Format Lyfo Disk
Manufacturing Origin United States
Product Type Quality Control
For Use With (Application) Antimicrobial Susceptibility Testing, Microbial Identification
Packaging Type Plastic Canister
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.