Learn More
ATCC Klebsiella pneumoniae; Strain 1100975
Catalog No. 502384281
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain 1100975 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is resistant to ertapenem, doripenem, imipenem, and meropenem and was confirmed by PCR to contain the blaNDM gene. The strain can be used in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.
Catalog No. 50-238-4281 Supplier ATCC Supplier No. BAA2472
Klebsiella pneumoniae subsp. pneumoniae strain 1100975 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is resistant to ertapenem, doripenem, imipenem, and meropenem and was confirmed by PCR to contain the blaNDM gene. The strain can be used in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.