Learn More
ATCC Klebsiella pneumoniae; Strain 1101160
Catalog No. 502384957
Encompass
Klebsiella pneumoniae strain 1101160 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This is a KPC reference strain that is resistant to ertapenem and imipenem. The strain has applications in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.
Catalog No. 50-238-4957 Supplier ATCC Supplier No. BAA2342
Klebsiella pneumoniae strain 1101160 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This is a KPC reference strain that is resistant to ertapenem and imipenem. The strain has applications in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.