Learn More
ATCC Klebsiella pneumoniae; Strain 1002565
Catalog No. 502382821
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain 1002565 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is resistant to ertapenem and imipenem and was confirmed by PCR to contain the blaNDM gene. This bacterial culture can be used in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.
Catalog No. 50-238-2821 Supplier ATCC Supplier No. BAA2470
Klebsiella pneumoniae subsp. pneumoniae strain 1002565 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is resistant to ertapenem and imipenem and was confirmed by PCR to contain the blaNDM gene. This bacterial culture can be used in antimicrobial resistance research, drug development, infectious disease research, and respiratory disease research.