Learn More
ATCC Klebsiella pneumoniae; Strain ART 2008133
Catalog No. 502381755
Encompass
Klebsiella pneumoniae strain ART 2008133 [D-05, 1338] was isolated from urine. This strain is a Klebsiella pneumoniae carbapenemase (KPC) producer and is used in in the Modified Hodge Test (MHT) as a positive control.
Catalog No. 50-238-1755 Supplier ATCC Supplier No. BAA1705
Klebsiella pneumoniae strain ART 2008133 [D-05, 1338] was isolated from urine. This strain is a Klebsiella pneumoniae carbapenemase (KPC) producer and is used in in the Modified Hodge Test (MHT) as a positive control.