Learn More
ATCC Klebsiella pneumoniae; Strain AIS2007023
Supplier:  ATCC BAA1706
Encompass
Klebsiella pneumoniae strain AIS 2007023 [6179] was confirmed by PCR to be negative for the blaKPC gene. This strain can be used in the Modified Hodge Test (HMT) as a negative control.
Catalog No. 50-238-1904