Learn More
ATCC Klebsiella pneumoniae; Strain: KP1074
Catalog No. 502383261
Encompass
Klebsiella pneumoniae strain KP1074 [B21] is a whole-genome sequenced bacterium that was isolated from a human with leukemia/lymphoma in Tel-Hashomer, Israel. This strain produces Klebsiella pneumoniae carbapenemase (KPC-3). The depositor states that this strain can be used as a QC reference strain for testing new beta-lactam/beta-lactamase inhibitor combinations such as meropenem/vaborbactam. The bacterium is a CLSI control strain for antimicrobial susceptibility testing.
Catalog No. 50-238-3261 Supplier ATCC Supplier No. BAA2814
Klebsiella pneumoniae strain KP1074 [B21] is a whole-genome sequenced bacterium that was isolated from a human with leukemia/lymphoma in Tel-Hashomer, Israel. This strain produces Klebsiella pneumoniae carbapenemase (KPC-3). The depositor states that this strain can be used as a QC reference strain for testing new beta-lactam/beta-lactamase inhibitor combinations such as meropenem/vaborbactam. The bacterium is a CLSI control strain for antimicrobial susceptibility testing.