Learn More
ATCC Klebsiella pneumoniae subsp. pneumoniae (Schroeter) Trevisan
Supplier:  ATCC 700603/DEL
Encompass
Control for extended-spectrum beta-lactamase production. Produces beta-lactamase SHV-18. This organism is a CLSI quality control strain for antimicrobial susceptibility testing
Catalog No. 50-189-185FP