Learn More
ATCC Klebsiella pneumoniae; subsp. pneumoniae
Supplier:  ATCC 35657
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain LRA 24 11 73 is a whole-genome sequenced strain. The strain can be used in infectious disease research, quality control testing, and respiratory disease research.
Catalog No. 50-238-2782
 Restricted Item