Learn More
ATCC Klebsiella pneumoniae; subsp. pneumoniae
Supplier:  ATCC 13883
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain NCTC 9633 is a bacterial type strain with applications in antimicrobial preservative testing, bioresistance testing, media testing, and quality control.
Catalog No. 50-238-4087