Learn More
ATCC Klebsiella pneumoniae; subsp. pneumoniae
Catalog No. 502384419
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain [CIP 104216, NCIB 10341] was isolated from cow's milk. This whole-genome sequenced bacterial strain can be used in inhibition testing, disinfectant testing, quality control, and respiratory disease research.
Catalog No. 50-238-4419 Supplier ATCC Supplier No. 4352
Klebsiella pneumoniae subsp. pneumoniae strain [CIP 104216, NCIB 10341] was isolated from cow's milk. This whole-genome sequenced bacterial strain can be used in inhibition testing, disinfectant testing, quality control, and respiratory disease research.