Learn More
ATCC Klebsiella pneumoniae; subsp. pneumoniae
Supplier:  ATCC 33495
Encompass
Klebsiella pneumoniae subsp. pneumoniae strain C122 is a whole-genome sequenced strain that was isolated from a human urinary tract. This bacterial culture can be used in infectious disease research, quality control, and respiratory disease research.
Catalog No. 50-238-3637