Learn More
ATCC Neisseria gonorrhoeae
Supplier:  ATCC 700719
Encompass
Resistant to spectinomycin [actinospectacin]. Quality control organism for Difco spectinomycin differentiation disks.
Catalog No. 50-238-0457