Learn More
ATCC Klebsiella pneumoniae; subsp. pneumoniae; Strain 1103199
Catalog No. 502382797
Encompass
Klebsiella pneumoniae subsp. pneumoniae is a clinical isolate from BioMerieux that can be used as a quality control strain for chromID Oxa 48 medium. This strain is resistant to ertapenem, imipenem, and meropenem and the depositor claims that this strain is a carbapenemase producer (OXA-48).
Catalog No. 50-238-2797 Supplier ATCC Supplier No. BAA2524
Klebsiella pneumoniae subsp. pneumoniae is a clinical isolate from BioMerieux that can be used as a quality control strain for chromID Oxa 48 medium. This strain is resistant to ertapenem, imipenem, and meropenem and the depositor claims that this strain is a carbapenemase producer (OXA-48).