Learn More
ATCC Pseudomonas sp.
Supplier: ATCC 31483
Pseudomonas sp. strain 3P is a bacterium that was isolated in South Carolina from a wastewater lagoon. This strain is cited to degrade detergents and surface active agents (US Patent 4,274,954, US Patent 4,317,885, US Patent 4,483,923).