Learn More
MSC KleenGuard Coverall, Liquid Particle Protection, Antistatic, Isocyanate Chromate Protection, 25/Pk.
Catalog No. 5000675231
Encompass
KleenGuard A45 Liquid & Particle Protection Surface Prep & Paint Apparel is designed for use in surface preparation and paint application areas, particularly where protection against isocyanate and chromate dust is mandated. This coverall features a lighter weight fabric, enhancing comfort without compromising protection, and is engineered for increased freedom of movement and reduced restriction.
  • Silicone free
  • Passes NFPA 99 for antistatic materials
  • Protects against isocyanates
  • Protects against chromate dust
  • Respirator fit hood design
  • Water resistant zipper
  • Storm flap
  • Elastic back
Catalog No. 50-006-75231 Supplier MSC Supplier No. 44629715
KleenGuard A45 Liquid & Particle Protection Surface Prep & Paint Apparel is designed for use in surface preparation and paint application areas, particularly where protection against isocyanate and chromate dust is mandated. This coverall features a lighter weight fabric, enhancing comfort without compromising protection, and is engineered for increased freedom of movement and reduced restriction.
  • Silicone free
  • Passes NFPA 99 for antistatic materials
  • Protects against isocyanates
  • Protects against chromate dust
  • Respirator fit hood design
  • Water resistant zipper
  • Storm flap
  • Elastic back