Learn More
MSC KleenGuard Coveralls, Liquid and Particle Protection, Large, Microporous Film, 25/Case
Catalog No. 5000769680
Encompass
KleenGuard A40 Liquid and Particle Protection Coveralls are constructed from microporous film laminate with a hood, zipper front, and elastic wrists and ankles, designed for protection against liquid splashes and dry particulates.
  • Made with blue breathable back
  • Offers superior liquid and particulate barrier compared to Tyvek
  • Features zipper closure, serged seams, and elastic cuffs and ankles
  • Passes NFPA 99 criteria for antistatic materials
  • Meets ASTM F1670 and ANSI/ISEA 101-1996 standards
  • Non-cleanroom ISO class
  • Supplied as 25 white garments per case
Catalog No. 50-007-69680 Supplier MSC Supplier No. 46813101
KleenGuard A40 Liquid and Particle Protection Coveralls are constructed from microporous film laminate with a hood, zipper front, and elastic wrists and ankles, designed for protection against liquid splashes and dry particulates.
  • Made with blue breathable back
  • Offers superior liquid and particulate barrier compared to Tyvek
  • Features zipper closure, serged seams, and elastic cuffs and ankles
  • Passes NFPA 99 criteria for antistatic materials
  • Meets ASTM F1670 and ANSI/ISEA 101-1996 standards
  • Non-cleanroom ISO class
  • Supplied as 25 white garments per case