Learn More
MSC KleenGuard Coveralls, Liquid and Particle Protection, Microporous Film, 2X-Large, 25/Case
Catalog No. 5000789291
Encompass
These A40 liquid and particle protection coveralls are constructed from a microporous film laminate, featuring a hood, zipper front, and elastic wrists and ankles. They provide superior liquid and particulate barrier protection, designed to shield against debris, dry particulates, and liquid splashes.
  • Made of microporous film laminate
  • Features a hood, zipper front, and elastic wrists and ankles
  • Offers protection against liquid and particulates
  • Designed to keep out debris, dry particulates, and liquid splashes
  • Passes NFPA 99 criteria for antistatic materials
  • Meets ASTM F1670 and ANSI/ISEA 101-1996 standards
  • Serged seams for enhanced durability
  • Supplied as 25 white garments per case
Catalog No. 50-007-89291 Supplier MSC Supplier No. 46813085
These A40 liquid and particle protection coveralls are constructed from a microporous film laminate, featuring a hood, zipper front, and elastic wrists and ankles. They provide superior liquid and particulate barrier protection, designed to shield against debris, dry particulates, and liquid splashes.
  • Made of microporous film laminate
  • Features a hood, zipper front, and elastic wrists and ankles
  • Offers protection against liquid and particulates
  • Designed to keep out debris, dry particulates, and liquid splashes
  • Passes NFPA 99 criteria for antistatic materials
  • Meets ASTM F1670 and ANSI/ISEA 101-1996 standards
  • Serged seams for enhanced durability
  • Supplied as 25 white garments per case