Learn More
MSC KleenGuard Disposable Coveralls, Film Laminate, 2X-Large, Non-Hazardous Protection, 1 ea
Catalog No. 5000792763
Encompass
These disposable coveralls are designed to provide non-hazardous protection. Made from a film laminate material, they feature a zipper closure, serged seams, and open cuffs and ankles. These coveralls are 2X-Large and are suitable for non-cleanroom ISO class environments.
  • Designed for non-hazardous protection
  • Constructed from film laminate material
  • Features a convenient zipper closure
  • Serged seams enhance durability
  • Open cuffs and open ankles for comfort
  • White color
  • Suitable for non-cleanroom ISO class environments
Catalog No. 50-007-92763 Supplier MSC Supplier No. 44629723
These disposable coveralls are designed to provide non-hazardous protection. Made from a film laminate material, they feature a zipper closure, serged seams, and open cuffs and ankles. These coveralls are 2X-Large and are suitable for non-cleanroom ISO class environments.
  • Designed for non-hazardous protection
  • Constructed from film laminate material
  • Features a convenient zipper closure
  • Serged seams enhance durability
  • Open cuffs and open ankles for comfort
  • White color
  • Suitable for non-cleanroom ISO class environments