Learn More
MSC KleenGuard Disposable Coveralls, Liquid Particle Protection, Microporous Film, 25 Pk
Catalog No. 5000688337
Encompass
These disposable coveralls provide liquid and particle protection, featuring a lightweight microporous film material for defense against hazardous liquids and dry particles. Designed for various industrial and manufacturing applications, they are suitable for environments requiring personal protective equipment.
  • Microporous film-laminated material
  • Protects against hazardous liquids (splashes) and dry particles (dust and dirt)
  • Features a hood, elastic wrists, and ankles
  • Zipper closure with serged seams
  • Non-Cleanroom ISO Class
  • Meets ANSI/ISEA 101-1996 standards
Catalog No. 50-006-88337 Supplier MSC Supplier No. 91435487
These disposable coveralls provide liquid and particle protection, featuring a lightweight microporous film material for defense against hazardous liquids and dry particles. Designed for various industrial and manufacturing applications, they are suitable for environments requiring personal protective equipment.
  • Microporous film-laminated material
  • Protects against hazardous liquids (splashes) and dry particles (dust and dirt)
  • Features a hood, elastic wrists, and ankles
  • Zipper closure with serged seams
  • Non-Cleanroom ISO Class
  • Meets ANSI/ISEA 101-1996 standards